Få mere at vide
N-Hexadecanol-D31, TRC
Artikelnummer. 30408536
Skift visning
Mængde:
20 mg
Artikelnummer. Mængde
30408536 20 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408536 Leverandør Toronto Research Chemicals Leverandørnr. TRCH29391820MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Hexadecanol-D31
CAS 203633-15-2
Molekylær formel C16 D31 H3 O
Mængde 20 mg
Synonym 1-Hexadecan-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-d31-ol (9CI), n-Hexadecanol-D31, n-Hexadecyl-2,2,3,3...16,16,16-d31 alcohol, Cetyl alcohol-d31, Cetyl-d31 alcohol, 1-Hexadecanol-2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-D31
InChI formel InChI=1S/C16H34O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h17H,2-16H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])CO
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,16-hentriacontadeuteriohexadecan-1-ol
Molekylvægt (g/mol) 273.632
Formel vægt 273.456