Få mere at vide
n-Heptyl-d15 Alcohol, CDN
Artikelnummer. 30310338
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30310338 0.1 g 0.1g
30311487 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310338 Leverandør CDN Leverandørnr. CDND39880.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Heptanol-D15
CAS 194793-95-8
Molekylær formel C7 D15 H O
Mængde 0.1 g
Synonym 1-Heptan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-d15-ol (9 CI), n-Heptyl-d15 alcohol, 1-Heptan-d15-ol, Heptyl-1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-d15 alcohol, 1-Heptanol-D15
InChI formel InChI=1 S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecadeuterioheptan-1-ol
Molekylvægt (g/mol) 131.294
Formel vægt 131.214 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity