Få mere at vide
n-Heptyl-5,5,6,6,7,7,7-d7 Alcohol, CDN
Artikelnummer. 30310619
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310619 0.1 g
30313642 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310619 Leverandør CDN Leverandørnr. CDND69200.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-Heptanol-5,5,6,6,7,7,7-D7
CAS 1219804-99-5
Molekylær formel C7 D7 H9 O
Mængde 0.1 g
Synonym n-Heptyl-5,5,6,6,7,7,7-d7 alcohol, 1-Heptan-5,5,6,6,7,7,7-d7-ol, Heptyl-5,5,6,6,7,7,7-d7 alcohol, 1-Heptanol-5,5,6,6,7,7,7-D7
InChI formel InChI=1 S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3/i1D3,2D2,3D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])CCCCO
IUPAC navn 5,5,6,6,7,7,7-heptadeuterioheptan-1-ol
Molekylvægt (g/mol) 123.245
Formel vægt 123.164 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity