Få mere at vide
N-Glycylproline, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Glycyl-L-proline |
| CAS | 704-15-4 |
| Molekylær formel | C7H12N2O3 |
| Mængde | 1 g |
| Synonym | Glycyl-L-proline, L-Proline, 1-glycyl-, Proline, 1-glycyl-, L- (6CI,8CI), Glycyl-L-proline, (S)-1-(2-Aminoacetyl)pyrrolidine-2-carboxylic acid, 1-Glycylproline, 11: PN: CN101988058 SEQID: 11 claimed sequence, 12: PN: WO2014063098 TABLE: 5 claimed sequence, 132: PN: US20130123467 SEQID: 158 claimed protein, 152: PN: EP2161028 PAGE: 10 claimed protein, 15: PN: WO2015153838 PAGE: 89 claimed sequence, 2: PN: TWI417385 SEQID: 11 claimed sequence, 2: PN: WO2011150133 PAGE: 63 claimed sequence, 4: PN: WO2016023974 PAGE: 31 claimed sequence, 7: PN: WO03052099 PAGE: 83 claimed protein, 81: PN: WO2011146121 PAGE: 115 claimed sequence, N-Glycyl-L-proline, N-Glycylproline, NSC 97929 |
| InChI formel | InChI=1S/C7H12N2O3/c8-4-6(10)9-3-1-2-5(9)7(11)12/h5H,1-4,8H2,(H,11,12)/t5-/m0/s1 |
| SMIL | NCC(=O)N1CCC[C@H]1C(=O)O |
| IUPAC navn | (2S)-1-(2-aminoacetyl)pyrrolidine-2-carboxylic acid |
| Molekylvægt (g/mol) | 172.182 |
| Formel vægt | 172.0848 |