Få mere at vide
N-Formylphenylephrine, Mikromol™
Artikelnummer. 17954396
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17954396 100 mg
17931776 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17954396 Leverandør Mikromol Leverandørnr. MM0081.15
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
N-Formylphenylephrine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Formylphenylephrine
CAS 2382194-29-6
Analyt- eller komponentnavne N-Formylphenylephrine
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C10 H13 N O3
Certificeringer/compliance ISO 17025
Synonym 2-[2-Hydroxy-5-[(4-ethyl-1-piperazinyl)sulfonyl]phenyl]-5-methyl-7-propyl-imidazo[5,1-f][1,2,4]triazin-4(1H)-one, N-Formylphenylephrine, N-((2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl)-N-methylformamide
InChI formel InChI=1S/2C13H19NO3.H2O4S/c2*1-13(2,3)14-6-10-9(12(17)7-14)4-8(15)5-11(10)16;1-5(2,3)4/h2*4-5,12,15-17H,6-7H2,1-3H3;(H2,1,2,3,4)
SMIL CN(C[C@H](O)c1cccc(O)c1)C=O
IUPAC navn N-[(2R)-2-hydroxy-2-(3-hydroxyphenyl)ethyl]-N-methylformamide
Molekylvægt (g/mol) 195.22
Mængde 100 mg
Formel vægt 195.0895 g/mol