missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Formylindoline, TRC

Artikelnummer. 30401370
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30401370 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30401370 Leverandør Toronto Research Chemicals Leverandørnr. TRCF70053025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Formylindoline
CAS 2861-59-8
Molekylær formel C9H9NO
Mængde 25 mg
Synonym 2,3-Dihydro-1H-indole-1-carboxaldehyde, 1-Indolinecarboxaldehyde
InChI formel InChI=1S/C9H9NO/c11-7-10-6-5-8-3-1-2-4-9(8)10/h1-4,7H,5-6H2
SMIL O=CN1CCc2ccccc12
Molekylvægt (g/mol) 147.17
Formel vægt 147.0684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.