missing translation for 'onlineSavingsMsg'
Learn More

N-Formylglycine-13C2 Ethyl Ester, TRC

Product Code. 30388803
Change view
Click to view available options
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30388803 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388803 Supplier Toronto Research Chemicals Supplier No. TRCF70046225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale N-Formylglycine-13C2 Ethyl Ester
CAS 1391051-73-2
Molekylær formel C313C2H9NO3
Mængde 25 mg
Synonym Dephenethylleccinine A-13C2, Ethyl N-formylglycinate-13C2, Ethyl formamidoacetate-13C2, Ethyl formylaminoacetate-13C2, NSC 14440-13C2
InChI formel InChI=1S/C5H9NO3/c1-2-9-5(8)3-6-4-7/h4H,2-3H2,1H3,(H,6,7)/i3+1,5+1
SMIL CCO[13C](=O)[13CH2]NC=O
IUPAC navn ethyl 2-formamidoacetate
Molekylvægt (g/mol) 133.12
Formel vægt 133.065
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.