Få mere at vide
N-Formyl Erythromycin A (>85%), TRC
Artikelnummer. 30138656
Skift visning
Mængde:
2.5 mg
10 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30138656 25 mg -
17913835 2.5 mg 2.5mg
17974405 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138656 Leverandør Toronto Research Chemicals Leverandørnr. TRCF70103025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 127955-44-6
Molekylær formel C37 H65 N O14
Molekylvægt (g/mol) 747.91
Synonym Erythromycin, N-demethyl-N-formyl-, N-Demethyl-N-formylerythromycin, Oxacyclotetradecane, erythromycin deriv., (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-4-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-14-ethyl-7,12,13-trihydroxy-3,5,7,9,11,13-hexamethyl-6-[[3,4,6-trideoxy-3-(formylmethylamino)-β-D-xylo-hexopyranosyl]oxy]oxacyclotetradecane-2,10-dione, 3' '-N-Demethyl-3' '-N-formylerythromycin A
IUPAC navn N-[(2S,3R,4S,6R)-2-[[(3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-14-ethyl-7,12,13-trihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,7,9,11,13-hexamethyl-2,10-dioxo-oxacyclotetradec-6-yl]oxy]-3-hydroxy-6-methyloxan-4-yl]-N-methylformamide
SMIL CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C=O)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O
Tekniske data
Kemisk navn eller materiale N-Formyl Erythromycin A
Mængde 25 mg
InChI formel InChI=1S/C37H65NO14/c1-13-25-37(10,46)30(42)20(4)27(40)18(2)15-35(8,45)32(52-34-28(41)24(38(11)17-39)14-19(3)48-34)21(5)29(22(6)33(44)50-25)51-26-16-36(9,47-12)31(43)23(7)49-26/h17-26,28-32,34,41-43,45-46H,13-16H2,1-12H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1
Formel vægt 747.4405