missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-L-alanine, 95%

Brand:  Alfa Aesar™ H63411.06

Additional Details : CAS Number : 84000-07-7 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470089

  • 539.56 DKK / 5g


In Stock 5
Add to basket

Description

Description

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

84000-07-7
325.364
JOFHWKQIQLPZTC-LBPRGKRZSA-N
688634
CC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C19H19NO4
MFCD00153384
Fmoc-N-Me-Ala-OH
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]propanoic acid

Specifications

N-Fmoc-N-methyl-L-alanine
95%
MFCD00153384
Fmoc-N-Me-Ala-OH
CC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
325.364
325.36
140°C
84000-07-7
C19H19NO4
4329998
JOFHWKQIQLPZTC-LBPRGKRZSA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]propanoic acid
688634
Solid
5g
Videos
SDS