missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-N-methyl-L-alanine, 95%

Brand:  Alfa Aesar™ H63411.14

Additional Details : CAS Number : 84000-07-7 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15480089

  • 1819.74 DKK / 25g


In stock, ready to ship: 2
Add to basket

Description

Description

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Also used in N-methyl amino acids for peptide synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

84000-07-7
325.364
JOFHWKQIQLPZTC-LBPRGKRZSA-N
688634
CC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C19H19NO4
MFCD00153384
fmoc-n-me-ala-oh, fmoc-n-methyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino propanoic acid, fmoc-nalpha-methyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-alanine, fmoc-l-meala-oh, ambotzfaa1395
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]propanoic acid

Specifications

N-Fmoc-N-methyl-L-alanine
C19H19NO4
4329998
Slightly soluble in water.
CC(C(=O)O)N(C)C(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
325.364
325.36
95%
25g
84000-07-7
MFCD00153384
fmoc-n-me-ala-oh, fmoc-n-methyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino propanoic acid, fmoc-nalpha-methyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino propanoic acid, l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxycarbonyl methyl amino propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-alanine, fmoc-l-meala-oh, ambotzfaa1395
JOFHWKQIQLPZTC-LBPRGKRZSA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]propanoic acid
688634
Solid
140°C
Videos
SDS Documents