missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-methionine sulfoxide, 98%, Thermo Scientific Chemicals

Catalog Number p-7054848
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
76265-70-8
C20H21NO5S
387.45
MFCD00077064
CEHRSUBRZOGRSW-HSYKDVHTSA-N
fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
7408215
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid
CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
76265-70-8
C20H21NO5S
387.45
MFCD00077064
CEHRSUBRZOGRSW-HSYKDVHTSA-N
fmoc-met o-oh, fmoc-l-methionine sulfoxide, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylsulfinyl butanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methanesulfinylbutanoic acid, ambotzfaa1405, n-9-fluorenylmethoxycarbonyl-methionine sulfoxide, n-alpha-9-fluorenylmethyloxycarbonyl-l-methioninesulfoxid
7408215
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylsulfinylbutanoic acid
CS(=O)CC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 387.45
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material N-Fmoc-L-methionine sulfoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'