Learn More
Alfa Aesar™ N-Fmoc-L-glutamic acid 1-tert-butyl ester, 98%
Brand: Alfa Aesar™ H62146.03
Additional Details : CAS Number : 84793-07-7 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
84793-07-7 | |
425.481 | |
GOPWHXPXSPIIQZ-FQEVSTJZSA-N | |
7017912 | |
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 |
C24H27NO6 | |
MFCD00065648 | |
fmoc-glu-otbu, fmoc-l-glutamic acid 1-tert-butyl ester, fmoc-glu obut-oh, fmoc-glu-obut, 4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, fmoc-glu-ot-bu, fmoc-glu-o-t-bu, pubchem10014, ksc448a0b | |
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid |
Specifications
N-Fmoc-L-glutamic acid 1-tert-butyl ester | |
84793-07-7 | |
MFCD00065648 | |
GOPWHXPXSPIIQZ-FQEVSTJZSA-N | |
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid | |
7017912 | |
98% |
1g | |
C24H27NO6 | |
fmoc-glu-otbu, fmoc-l-glutamic acid 1-tert-butyl ester, fmoc-glu obut-oh, fmoc-glu-obut, 4s-5-tert-butoxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-tert-butoxy-5-oxopentanoic acid, fmoc-glu-ot-bu, fmoc-glu-o-t-bu, pubchem10014, ksc448a0b | |
CC(C)(C)OC(=O)C(CCC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13 | |
425.481 | |
425.47 |