missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-L-glutamic acid 1-benzyl ester, 95%

Brand:  Alfa Aesar™ H66551.03

Additional Details : CAS Number : 122350-52-1 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15482779

  • 290.11 DKK / 1g


In stock, ready to ship: 1
Add to basket

Specifications

Chemical Identifiers

122350-52-1
459.498
FMWLYDDRYGOYMY-DEOSSOPVSA-N
10049854
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C27H25NO6
MFCD00080272
fmoc-glu-obzl, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 4s-5-benzyloxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, fmoc-l-glutamic acid alpha-benzyl ester, nalpha-fmoc-l-glutamic acid alpha-benzyl ester
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid

Specifications

N-Fmoc-L-glutamic acid 1-benzyl ester
C27H25NO6
fmoc-glu-obzl, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, 4s-5-benzyloxy-4-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, fmoc-l-glutamic acid alpha-benzyl ester, nalpha-fmoc-l-glutamic acid alpha-benzyl ester
C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
459.498
459.5
95%
1g
122350-52-1
MFCD00080272
FMWLYDDRYGOYMY-DEOSSOPVSA-N
(4S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid
10049854
Powder
White
Videos
SDS Documents