missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-citrulline, 98%, Thermo Scientific Chemicals

Catalog Number p-7054905
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
133174-15-9
C21H23N3O5
397.431
MFCD00151943
NBMSMZSRTIOFOK-SFHVURJKSA-N
fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002
2756127
(2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O
This item is not returnable. View return policy
133174-15-9
C21H23N3O5
397.431
MFCD00151943
NBMSMZSRTIOFOK-SFHVURJKSA-N
fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002
2756127
(2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 7780987
Formula Weight 397.42
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-citrulline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'