missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Quantity | 250 mg |
Beilstein | 7726557 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 401.46 |
Percent Purity | 95% |
Chemical Name or Material | N-Fmoc-L-beta-homophenylalanine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'