missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Fmoc-4-iodo-D-phenylalanine, 98%

Brand:  Alfa Aesar™ H63292.06

Additional Details : CAS Number : 205526-29-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469918

  • 2080.30 DKK / 5g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Description

Description

Applications
N-Fmoc-4-iodo-D-phenylalanine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly Soluble in water (6.7E-4 g/L) (25°C).

Notes
Light sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

205526-29-0
513.331
LXOXXTQKKRJNNB-JOCHJYFZSA-N
16213204
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)I)C(=O)O
C24H20INO4
MFCD00672557
fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid

Specifications

N-Fmoc-4-iodo-D-phenylalanine
C24H20INO4
9159551
LXOXXTQKKRJNNB-JOCHJYFZSA-N
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid
16213204
Solid
5g
205526-29-0
MFCD00672557
fmoc-d-phe 4-i-oh, fmoc-4-iodo-d-phenylalanine, fmoc-d-4-iodophenylalanine, fmoc-d-4-iodophe, fmoc-4'-iodo-d-phe, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-iodophenyl propanoic acid, ambotzfaa1584, fmoc-4-iodo-d-phe-oh, fmoc-d-phe 4-i-oh hplc
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)I)C(=O)O
513.331
513.32
98%
Light Sensitive
Videos
SDS