missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(1-naphthyl)-D-alanine, 98%, Thermo Scientific Chemicals

Catalog Number 15480509
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
138774-93-3
C28H23NO4
437.50
MFCD00191200
ORWNVJDLEMVDLV-UHFFFAOYNA-N
fmoc-3-1-naphthyl-d-alanine, fmoc-d-1-nal-oh, fmoc-d-1-naphthylalanine, fmoc-beta-1-naphthyl-d-alanine, fmoc-d-3-1-naphthyl-alanine, fmoc-3-1-naphthyl-d-ala, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-naphthalen-1-yl-propionic acid, n-fmoc-3-1-naphthyl-l-alanine, ambotzfaa1659
7129831
OC(=O)C(CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
138774-93-3
C28H23NO4
437.50
MFCD00191200
ORWNVJDLEMVDLV-UHFFFAOYNA-N
fmoc-3-1-naphthyl-d-alanine, fmoc-d-1-nal-oh, fmoc-d-1-naphthylalanine, fmoc-beta-1-naphthyl-d-alanine, fmoc-d-3-1-naphthyl-alanine, fmoc-3-1-naphthyl-d-ala, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-naphthalen-1-yl-propionic acid, n-fmoc-3-1-naphthyl-l-alanine, ambotzfaa1659
7129831
OC(=O)C(CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 9160026
IUPAC Name 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-1-yl)propanoic acid
Formula Weight 437.49
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Fmoc-3-(1-naphthyl)-D-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'