Få mere at vide
N-Ethylsuccinimide-d5, TRC
Artikelnummer. 30375822
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30375822 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30375822 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92596250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Ethylsuccinimide-d5
CAS 1370698-98-8
Molekylær formel C6H4D5NO2
Mængde 50 mg
Synonym 1-(Ethyl-1,1,2,2,2-d5)-2,5-pyrrolidinedione, NSC 38693-d5
InChI formel InChI=1S/C6H9NO2/c1-2-7-5(8)3-4-6(7)9/h2-4H2,1H3/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])N1C(=O)CCC1=O
IUPAC navn 1-(1,1,2,2,2-pentadeuterioethyl)pyrrolidine-2,5-dione
Molekylvægt (g/mol) 132.17
Formel vægt 132.0947