Få mere at vide
N-Ethylpiperazine-d5 Bis(trifluoroacetic Acid) Salt, TRC
Artikelnummer. 30369952
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30369952 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30369952 Leverandør Toronto Research Chemicals Leverandørnr. TRCE92549710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Ethylpiperazine-d5 Bis(trifluoroacetic Acid) Salt
CAS 1246817-22-0
Molekylær formel C10H11D5F6N2O4
Mængde 10 mg
Synonym 1-Ethyl-piperazine-d5 Bis(trifluoroacetic Acid) Salt, 1-Ethylpiperazine-d5 Bis(trifluoroacetic Acid) Salt
SMIL [2H]C([2H])(C([2H])([2H])[2H])N1CCNCC1.OC(C(F)(F)F)=O.OC(C(F)(F)F)=O
Molekylvægt (g/mol) 347.28