Få mere at vide
N-Ethyl-N-methylcarbamoyl Chlorid, TRC
Artikelnummer. 30008739
Skift visning
Mængde:
100 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008739 1 g
30008364 10 g
30006023 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008739 Leverandør Toronto Research Chemicals Leverandørnr. TRCE9251451G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 42252-34-6
Molekylær formel C4 H8 Cl N O
Molekylvægt (g/mol) 121.57
Synonym Bosutinib Impurity 41
IUPAC navn N-ethyl-N-methylcarbamoyl chloride
SMIL CCN(C)C(=O)Cl
Tekniske data
Kemisk navn eller materiale N-Ethyl-N-methylcarbamoyl Chloride
InChI formel InChI=1S/C4H8ClNO/c1-3-6(2)4(5)7/h3H2,1-2H3
Mængde 1 g
Formel vægt 121.0294