missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N-Ethyl-N-methyl-2-(1-piperazinyl)nicotinamide
Click to view available options
Quantity:
250 mg
1 g
Specifications
Quantity | 250 mg |
Formula Weight | 248.33 |
Chemical Name or Material | N-Ethyl-N-methyl-2-(1-piperazinyl)nicotinamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'