Få mere at vide
N-Ethyl-d5-maleimide, CDN
Artikelnummer. 30317936
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30317936 0.05 g
30323656 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317936 Leverandør CDN Leverandørnr. CDND61410.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Ethylmaleimide-D5 (N-ethyl-D5)
CAS 360768-37-2
Molekylær formel C6H2D5NO2
Mængde 0.05 g
Synonym 1 H-Pyrrole-2,5-dione, 1-(ethyl-d5)- (9 CI, ACI), 1-(Ethyl-d5)-1 H-pyrrole-2,5-dione (ACI), N-Ethylmaleimide-D5 (N-ethyl-D5), N-Ethylmaleimide-d5, N-(Ethyl-d5) maleimide
InChI formel InChI=1 S/C6H7NO2/c1-2-7-5(8)3-4-6(7)9/h3-4 H,2H2,1H3/i1D3,2D2
SMIL C(C([2 H])([2 H])[2 H])(N1C(=O)C=CC1=O)([2 H])[2 H]
IUPAC navn 1-(ethyl-d5)-1 H-pyrrole-2,5-dione
Molekylvægt (g/mol) 130.16
Formel vægt 130.08 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity