Få mere at vide
N-Ethyl-3-phenylpropan-1-amine Hydrochloride, Mikromol™
Beskrivelse
N-Ethyl-3-phenylpropan-1-amine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Ethyl-3-phenylpropan-1-amine Hydrochloride |
| CAS | 13125-63-8 |
| Analyt- eller komponentnavne | N-Ethyl-3-phenylpropan-1-amine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H17 N . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Alverine Citrate Imp. C (EP), Alverine Imp. C (EP), N-Ethyl-3-phenylpropan-1-amine Hydrochloride, Alverine Citrate Imp. C (EP) as Hydrochloride, Alverine Citrate Impurity C as Hydrochloride, Alverine Impurity C as Hydrochloride |
| InChI formel | InChI=1S/C11H17N.ClH/c1-2-12-10-6-9-11-7-4-3-5-8-11;/h3-5,7-8,12H,2,6,9-10H2,1H3;1H |
| SMIL | Cl.CCNCCCc1ccccc1 |
| IUPAC navn | N-ethyl-3-phenylpropan-1-amine;hydrochloride |
| Vis mere |