Få mere at vide
N-(Ethoxycarbonylmethyl)piperazine, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-(Ethoxycarbonylmethyl)piperazine |
| CAS | 40004-08-8 |
| Molekylær formel | C8H16N2O2 |
| Mængde | 100 mg |
| Synonym | 1-Piperazineacetic Acid Ethyl Ester, 2-(Piperazin-1-yl)acetic Acid Ethyl Ester, 4-(Carboethoxymethyl)piperazine, Ethyl 1-piperazineacetate, Ethyl 1-Piperazinylacetate, Ethyl 2-(piperazin-1-yl)acetate, Ethyl 2-Piperazinoacetate, Ethyl Piperazinoacetate, N-(Carboethoxymethyl)piperazine, N-(Ethoxycarbonylmethyl)piperazine, Piperazin-1-ylacetic Acid Ethyl Ester |
| InChI formel | InChI=1S/C8H16N2O2/c1-2-12-8(11)7-10-5-3-9-4-6-10/h9H,2-7H2,1H3 |
| SMIL | CCOC(=O)CN1CCNCC1 |
| Molekylvægt (g/mol) | 172.22 |
| Formel vægt | 172.1212 |