Få mere at vide
N-(Ethoxycarbonyl)-Alpha-methyl-L-tryptophan Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | rac-N-(ethoxycarbonyl)-alpha-methyl-L-tryptophan Methyl Ester |
| CAS | 1798895-24-5 |
| Molekylær formel | C16 H20 N2 O4 |
| Mængde | 500 mg |
| Synonym | N-(Ethoxycarbonyl)-alpha-methyl-L-tryptophan Methyl Ester, N-(Ethoxycarbonyl)-α-methyl-L-tryptophan Methyl Ester |
| InChI formel | InChI=1S/C16H20N2O4/c1-4-22-15(20)18-16(2,14(19)21-3)9-11-10-17-13-8-6-5-7-12(11)13/h5-8,10,17H,4,9H2,1-3H3,(H,18,20)/t16-/m1/s1 |
| SMIL | CCOC(=O)N[C@@](C)(Cc1c[nH]c2ccccc12)C(=O)OC |
| IUPAC navn | rac-(1E)-1-ethoxy-N-[rac-(1R)-1-(1H-indol-3-ylmethyl)-2-methoxy-1-methyl-2-oxo-ethyl]methanimidic acid |
| Molekylvægt (g/mol) | 304.34 |