Få mere at vide
n-Eicosanyl-d41 Alcohol, TRC
Artikelnummer. 30378465
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30378465 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30378465 Leverandør Toronto Research Chemicals Leverandørnr. TRCE5010061MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Eicosanyl-d41 Alcohol
CAS 349553-89-5
Molekylær formel C20 D41 H O
Mængde 1 mg
Synonym 1-Eicosan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20,20-d41-ol (9CI), 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20,20-Hentetracontadeuterioicosan-1-ol, n-Eicosanyl-d41 Alcohol, 1-Eicosanol-D41, Arachidyl-d41 alcohol, Arachic-d41 alcohol, Arachidic-d41 alcohol
InChI formel InChI=1S/C20H42O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21/h21H,2-20H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,15,15,16,16,17,17,18,18,19,19,20,20,20-hentetracontadeuterioicosan-1-ol
Molekylvægt (g/mol) 339.7995
Formel vægt 339.5809