Få mere at vide
N-(E)-3-Tributyltinallyl-1-naphthalene-d7-methylamine, TRC
Artikelnummer. 30451970
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30451970 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451970 Leverandør Toronto Research Chemicals Leverandørnr. TRCT77402210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-(E)-3-Tributyltinallyl-1-naphthalene-d7-methylamine
CAS 1310383-84-6
Molekylær formel C26 2H7 H34 N Sn
Mængde 10 mg
InChI formel InChI=1S/C14H14N.3C4H9.Sn/c1-3-11-15(2)14-10-6-8-12-7-4-5-9-13(12)14;3*1-3-4-2;/h1,3-10H,11H2,2H3;3*1,3-4H2,2H3;/i4D,5D,6D,7D,8D,9D,10D;;;;
SMIL [2H]c1c([2H])c([2H])c2c(N(C)C\C=C\[Sn](CCCC)(CCCC)CCCC)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn 2,3,4,5,6,7,8-heptadeuterio-N-methyl-N-[(E)-3-tributylstannylprop-2-enyl]naphthalen-1-amine
Molekylvægt (g/mol) 493.36
Formel vægt 494.27