Få mere at vide
N-Dodecyltrimethylenediamine, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Dodecyltrimethylenediamine |
| CAS | 5538-95-4 |
| Analyt- eller komponentnavne | N-Dodecyltrimethylenediamine |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C15 H34 N2 |
| Certificeringer/compliance | ISO 17034 |
| Synonym | N-Dodecyltrimethylenediamine, 1,3-Propanediamine, N1-dodecyl-, 1,3-Propanediamine, N-dodecyl- (6CI,7CI,8CI,9CI), N1-Dodecyl-1,3-propanediamine, 3-(Dodecylamino)-1-propanamine, 3-(Dodecylamino)propylamine, 3-(Laurylamino)propylamine, Diam 21, Dicrodamine 1.12, Genamin LAP 100, Genamin LAP 100D, Laurylaminopropylamine, N-(3-Aminopropyl)dodecylamine, N-Dodecyl-1,3-diaminopropane, N-Dodecyl-1,3-propanediamine, N-Dodecyl-1,3-propylenediamine, N-Dodecyl-1,3-trimethylenediamine, N-Dodecyltrimethylenediamine, N-Lauryl-1,3-diaminopropane, N-Lauryltrimethylenediamine, NSC 28753, NSC 40165, RC 5635 |
| InChI formel | InChI=1S/C15H34N2/c1-2-3-4-5-6-7-8-9-10-11-14-17-15-12-13-16/h17H,2-16H2,1H3 |
| SMIL | CCCCCCCCCCCCNCCCN |
| IUPAC navn | N'-dodecylpropane-1,3-diamine |
| Vis mere |