Få mere at vide
n-Dodecyltrimethylammonium-d34 Bromide, CDN
Artikelnummer. 30328490
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328490 0.05 g
30320601 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328490 Leverandør CDN Leverandørnr. CDND67280.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dodecyltrimethylammonium-D34 Bromide
CAS 2259752-12-8
Molekylær formel C15 D34 N . Br
Mængde 0.05 g
Synonym 1-Dodecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25-aminium, N,N,N-tri(methyl-d3)-, bromide (1:1) (ACI), Dodecyltrimethylammonium-d34 bromide, Dodecyl-d25-tri(methyl-d3)ammonium bromide, Lauryltrimethylammonium-d34 bromide, Dodecyltrimethylammonium bromide-d34, N,N,N-Tri(methyl-d3)-1-dodecanaminium-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d25 bromide
InChI formel InChI=1 S/C15H34N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2;
SMIL [Br-].[2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl-tris(trideuteriomethyl)azanium;bromide
Molekylvægt (g/mol) 342.5507
Formel vægt 341.4009 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity