Få mere at vide
n-Dodecylphosphocholine-d38, CDN
Artikelnummer. 30322283
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322283 0.1 g 0.1g
30321334 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322283 Leverandør CDN Leverandørnr. CDND36120.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Dodecylphosphocholine-D38
CAS 130890-78-7
Molekylær formel C17 D38 N O4 P
Mængde 0.1 g
Synonym Ethan-1,1,2,2-d4-aminium, 2-[[(dodecyl-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-tri(methyl-d3)-, inner salt (9 CI, ACI), n-Dodecylphosphocholine-d38, 2-[[(Dodecyl-d25-oxy)hydroxyphosphinyl]oxy]-N,N,N-tri(methyl-d3)ethan-1,1,2,2-d4-aminium inner salt
InChI formel InChI=1 S/C17H38NO4P/c1-5-6-7-8-9-10-11-12-13-14-16-21-23(19,20)22-17-15-18(2,3)4/h5-17H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])OP(=O)([O-])OC([2 H])([2 H])C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-pentacosadeuteriododecyl [1,1,2,2-tetradeuterio-2-[tris(trideuteriomethyl)azaniumyl]ethyl] phosphate
Molekylvægt (g/mol) 389.6958
Formel vægt 389.4924 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity