Få mere at vide
N-Dodecylacrylamide, 98+%, Thermo Scientific™
452.25 DKK - 1455.25 DKK
Kemiske identifikatorer
| CAS | 1506-53-2 |
|---|---|
| Molekylær formel | C15H29NO |
| Molekylvægt (g/mol) | 239.40 |
| MDL nummer | MFCD08276361 |
| InChI nøgle | XQPVIMDDIXCFFS-UHFFFAOYSA-N |
| Synonym | n-dodecylacrylamide, 2-propenamide,n-dodecyl, acmc-209d4r |
| PubChem CID | 73929 |
| IUPAC navn | N-dodecylprop-2-enamide |
| SMIL | CCCCCCCCCCCCNC(=O)C=C |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15447787
|
Thermo Scientific Alfa Aesar
H37507.06 |
5g |
452.25 DKK
5g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15457787
|
Thermo Scientific Alfa Aesar
H37507.14 |
25g |
1455.25 DKK
25g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Kemiske identifikatorer
| C15H29NO | |
| MFCD08276361 | |
| n-dodecylacrylamide, 2-propenamide,n-dodecyl, acmc-209d4r | |
| N-dodecylprop-2-enamide |
Tekniske data
| N-Dodecylacrylamide | |
| C15H29NO | |
| 4(4)804 | |
| XQPVIMDDIXCFFS-UHFFFAOYSA-N | |
| N-dodecylprop-2-enamide | |
| 73929 | |
| ≥98% |
| 1506-53-2 | |
| MFCD08276361 | |
| n-dodecylacrylamide, 2-propenamide,n-dodecyl, acmc-209d4r | |
| CCCCCCCCCCCCNC(=O)C=C | |
| 239.40 | |
| 239.4 |
Sikkerhed og håndtering
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.