missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyl-beta-D-maltopyranoside, Thermo Scientific Chemicals
690.59 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 mg
1 g
5 g
Description
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.
Solubility
Soluble in water at 10% w/v
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.
Solubility
Soluble in water at 10% w/v
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Melting Point | 224°C to 226°C |
Color | White |
Quantity | 500 mg |
Beilstein | 55318 |
Solubility Information | Soluble in water at 10% w/v |
Formula Weight | 510.62 |
Physical Form | Powder |
Chemical Name or Material | n-Dodecyl-beta-D-maltopyranoside |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'