Få mere at vide
n-Dodecane-d26, CDN
Artikelnummer. 30310818
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30310818 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310818 Leverandør CDN Leverandørnr. CDND08821G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Dodecane-d26
CAS 16416-30-1
Molekylær formel C12 D26
Mængde 1 g
Synonym Dodecane-1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-d26, Perdeuterododecane, Adakane 12-d26, Ba 51-090453-d26, C12-n-Alkane-d26, Dodecane-d26 (8 CI,9 CI), Dodecane-d26 (8 CI,9 CI), Dodecane-d26 (8 CI,9 CI), Dodecane-d26 (8 CI,9 CI), Dodecane-d26 (8 CI,9 CI), Dodecane-d26 (8 CI,9 CI), Dodecane-d26 (8 CI,9 CI)
InChI formel InChI=1 S/C12H26/c1-3-5-7-9-11-12-10-8-6-4-2/h3-12H2,1-2H3/i1D3,2D3,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-hexacosadeuteriododecane
Molekylvægt (g/mol) 196.5
Formel vægt 196.3666 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity