Få mere at vide
n-Dipropylamine-d4 Hydrochloride, TRC
Artikelnummer. 30377335
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30377335 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377335 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49215225MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale n-Dipropylamine-d4 Hydrochloride
CAS 1329834-71-0
Molekylær formel C6H12D4ClN
Mængde 25 mg
Synonym Di-n-propylamine-d4 Hydrochloride, Dipropanamine-d4 Hydrochlroide, N,N-Dipropylamine-d4 Hydrochloride, N-Propyl-1-propanamine-d4 Hydrochloride
InChI formel InChI=1S/C6H15N.ClH/c1-3-5-7-6-4-2;/h7H,3-6H2,1-2H3;1H/i5D2,6D2;
SMIL Cl.[2H]C([2H])(CC)NC([2H])([2H])CC
IUPAC navn 1,1-dideuterio-N-(1,1-dideuteriopropyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 141.68
Formel vægt 141.1222