Få mere at vide
N-Despropyl Ropinirole-d3, TRC
Artikelnummer. 30354763
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30354763 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30354763 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2973822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Despropyl Ropinirole-d3
CAS 1215798-00-7
Molekylær formel C13H15D3N2O
Mængde 2.5 mg
Synonym 4-[2-(Propyl-d3-amino)ethyl]-1,3-dihydro-2H-indol-2-one, 1,3-Dihydro-4-[2-(propyl-d3-amino)ethyl]-2H-indol-2-one, SKF 104557-d3
InChI formel InChI=1S/C13H18N2O/c1-2-7-14-8-6-10-4-3-5-12-11(10)9-13(16)15-12/h3-5,14H,2,6-9H2,1H3,(H,15,16)/i1D3
SMIL O=C(N1)CC2=C1C=CC=C2CCNCCC([2H])([2H])[2H]
Molekylvægt (g/mol) 221.31