missing translation for 'onlineSavingsMsg'
Få mere at vide

N-Desmethylvenlafaxine Hydrochloride, LoGiCal

Artikelnummer. 30458803
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458803 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30458803 Leverandør LoGiCal Leverandørnr. LGCFOR0393.04

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale N-Desmethylvenlafaxine Hydrochloride
CAS 93413-90-2
Analyt- eller komponentnavne N-Desmethylvenlafaxine Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C16 H25 N O2 . Cl H
Certificeringer/compliance ISO 9001
Synonym Venlafaxine Hydrochloride Imp. D (EP), Venlafaxine Imp. D (EP), Venlafaxine USP Related Compound A, Venlafaxine USP RC A, 1-[(1RS)-1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride, Venlafaxine Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethylvenlafaxine Hydrochloride, Venlafaxine Related Compound A as Hydrochloride, Venlafaxine Hydrochloride Impurity D as Hydrochloride, Venlafaxine Impurity D as Hydrochloride
InChI formel InChI=1S/C16H25NO2.ClH/c1-17-12-15(16(18)10-4-3-5-11-16)13-6-8-14(19-2)9-7-13;/h6-9,15,17-18H,3-5,10-12H2,1-2H3;1H
Løsningstype Neat
SMIL Cl.CNCC(c1ccc(OC)cc1)C2(O)CCCCC2
IUPAC navn 1-[1-(4-methoxyphenyl)-2-(methylamino)ethyl]cyclohexan-1-ol;hydrochloride
Industry Type Clinical Testing
Molekylvægt (g/mol) 299.84
Mængde 10 mg
Formel vægt 299.1652 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.