Få mere at vide
N-Desmethylvenlafaxine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desmethylvenlafaxine Hydrochloride |
| CAS | 93413-90-2 |
| Analyt- eller komponentnavne | N-Desmethylvenlafaxine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H25 N O2 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Venlafaxine Hydrochloride Imp. D (EP), Venlafaxine Imp. D (EP), Venlafaxine USP Related Compound A, Venlafaxine USP RC A, 1-[(1RS)-1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride, Venlafaxine Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethylvenlafaxine Hydrochloride, Venlafaxine Related Compound A as Hydrochloride, Venlafaxine Hydrochloride Impurity D as Hydrochloride, Venlafaxine Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C16H25NO2.ClH/c1-17-12-15(16(18)10-4-3-5-11-16)13-6-8-14(19-2)9-7-13;/h6-9,15,17-18H,3-5,10-12H2,1-2H3;1H |
| Løsningstype | Single Solution |
| SMIL | Cl.CNCC(c1ccc(OC)cc1)C2(O)CCCCC2 |
| Vis mere |