Få mere at vide
N-Desmethylolanzapine, Lipomed
Artikelnummer. 30458405
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30458405 50 mg
1 options
Artikelnummer. 30458405 Leverandør Lipomed Leverandørnr. LPMOLZ1899FB50
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
N-Desmethylolanzapine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Desmethylolanzapine
CAS 161696-76-0
Analyt- eller komponentnavne Desmethylolanzapine
Forsendelsestilstand Room Temperature
Molekylær formel C16 H18 N4 S
Synonym N-Desmethylolanzapine, Desmethylolanzapine, 2-Methyl-4-(1-piperazinyl)-10 H-thieno[2,3-b][1,5 ]benzodiazepine, 4'-Desmethylolanzapine, LY 170055, N-Demethylolanzapine
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMIL Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4
IUPAC navn 2-methyl-4-piperazin-1-yl-10 H-thieno[2,3-b][1,5 ]benzodiazepine
Industry Type Pharmaceutical
Molekylvægt (g/mol) 298.41
Mængde 50 mg
Formel vægt 298.1252 g/mol