missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
N-Desmethylolanzapine, Lipomed
Description
N-Desmethylolanzapine
Specifications
Specifications
| Kemisk navn eller materiale | Desmethylolanzapine |
| CAS | 161696-76-0 |
| Analyt- eller komponentnavne | Desmethylolanzapine |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H18 N4 S |
| Synonym | N-Desmethylolanzapine, Desmethylolanzapine, 2-Methyl-4-(1-piperazinyl)-10 H-thieno[2,3-b][1,5 ]benzodiazepine, 4'-Desmethylolanzapine, LY 170055, N-Demethylolanzapine |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMIL | Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4 |
| IUPAC navn | 2-methyl-4-piperazin-1-yl-10 H-thieno[2,3-b][1,5 ]benzodiazepine |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?