Få mere at vide
N-Desmethylolanzapine Dihydrochloride, Mikromol™
Beskrivelse
N-Desmethylolanzapine Dihydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-Desmethylolanzapine Dihydrochloride |
| CAS | 2319721-48-5 |
| Analyt- eller komponentnavne | N-Desmethylolanzapine Dihydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H18 N4 S . 2 Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Desmethylolanzapine Dihydrochloride |
| InChI formel | InChI=1S/C16H18N4S.2ClH/c1-11-10-12-15(20-8-6-17-7-9-20)18-13-4-2-3-5-14(13)19-16(12)21-11;;/h2-5,10,17,19H,6-9H2,1H3;2*1H |
| SMIL | Cl.Cl.Cc1cc2C(=Nc3ccccc3Nc2s1)N4CCNCC4 |
| IUPAC navn | 2-methyl-4-piperazin-1-yl-10H-thieno[2,3-b][1,5]benzodiazepine;hydrochloride |
| Vis mere |