Få mere at vide
N-Desmethylcyclobenzaprine Hydrochloride (Norcyclobenzaprine Hydrochloride) 1.0 mg/ml in Methanol (as free base), LoGiCal
Artikelnummer. 30462947
Skift visning
Mængde:
1 ml
Artikelnummer. Mængde
30462947 1 ml
1 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30462947 Leverandør LoGiCal Leverandørnr. LGCAMP0079.0101
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethylcyclobenzaprine HCl
CAS 438-59-5
Analyt- eller komponentnavne N-Desmethylcyclobenzaprine HCl
Forsendelsestilstand Ice pack (-20°C)
Molekylær formel C19 H19 N . Cl H
Certificeringer/compliance ISO 17034
Synonym Nortriptyline Hydrochloride Imp. B (EP), Nortriptyline Imp. B (EP), Cyclobenzaprine USP Related Compound B, Cyclobenzaprine USP RC B, 3-(5H-Dibenzo-[a,d][7]annulen-5-ylidene)-N-methylpropan-1-amine Hydrochloride, Nortriptyline Hydrochloride Imp. B (EP) as Hydrochloride, Norcyclobenzaprine Hydrochloride, Cyclobenzaprine Related Compound B as Hydrochloride, Nortriptyline Hydrochloride Impurity B as Hydrochloride, Nortriptyline Impurity B as Hydrochloride
InChI formel InChI=1S/C19H19N.ClH/c1-20-14-6-11-19-17-9-4-2-7-15(17)12-13-16-8-3-5-10-18(16)19;/h2-5,7-13,20H,6,14H2,1H3;1H
Løsningstype Single Solution
SMIL Cl.CNCCC=C1c2ccccc2C=Cc3ccccc13
IUPAC navn 3-(dibenzo[1,2-a:2',1'-d][7]annulen-11-ylidene)-N-methylpropan-1-amine;hydrochloride
Opløsningsmiddel eller Matrix Methanol
Industry Type Clinical Testing
Molekylvægt (g/mol) 297.82
Mængde 1 ml
Formel vægt 297.1284 g/mol