Få mere at vide
N-Desmethylcarboxy Terbinafine-d7, TRC
Artikelnummer. 30361735
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30361735 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361735 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2914071MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethylcarboxy Terbinafine-d7
CAS 1185238-58-7
Molekylær formel C20 D7 H14 N O2
Mængde 1 mg
Synonym (5E)-2,2-Dimethyl-7-[(1-naphthalenyl-d7-methyl)amino]-5-hepten-3-ynoic Acid
InChI formel InChI=1S/C20H21NO2/c1-20(2,19(22)23)13-6-3-7-14-21-15-17-11-8-10-16-9-4-5-12-18(16)17/h3-5,7-12,21H,14-15H2,1-2H3,(H,22,23)/b7-3+/i4D,5D,8D,9D,10D,11D,12D
SMIL [2H]c1c([2H])c([2H])c2c(CNC\C=C\C#CC(C)(C)C(=O)O)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn (E)-7-[(2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl)methylamino]-2,2-dimethylhept-5-en-3-ynoic acid
Molekylvægt (g/mol) 314.429
Formel vægt 314.201