Få mere at vide
N-Desmethylcarboxy Terbinafine-d7 Methyl Ester, TRC
Artikelnummer. 30355304
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30355304 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30355304 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2914122.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethylcarboxy Terbinafine-d7 Methyl Ester
CAS 1185245-14-0
Molekylær formel C21 2H7 H16 N O2
Mængde 2.5 mg
InChI formel InChI=1S/C21H23NO2/c1-21(2,20(23)24-3)14-7-4-8-15-22-16-18-12-9-11-17-10-5-6-13-19(17)18/h4-6,8-13,22H,15-16H2,1-3H3/b8-4+/i5D,6D,9D,10D,11D,12D,13D
SMIL [2H]c1c([2H])c([2H])c2c(CNC\C=C\C#CC(C)(C)C(=O)OC)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn methyl (E)-7-[(2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl)methylamino]-2,2-dimethylhept-5-en-3-ynoate
Molekylvægt (g/mol) 328.46
Formel vægt 328.2168