Få mere at vide
N-Desmethyl Vecuronium, TRC
Artikelnummer. 30366687
Skift visning
Mængde:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366687 5 mg
30366483 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30366687 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2964905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13529-31-2
Molekylær formel C33 H54 N2 O4
Molekylvægt (g/mol) 542.79
Synonym 2β,16β-Bis(piperidin-1-yl)-5α-androstane-3α,17β-diyl Diacetate
IUPAC navn [(2S,3S,5S,8R,9S,10S,13S,14S,16S,17R)-17-acetyloxy-10,13-dimethyl-2,16-di(piperidin-1-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMIL CC(=O)O[C@H]1C[C@@H]2CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3C[C@@H]([C@@H]4OC(=O)C)N5CCCCC5)[C@@]2(C)C[C@@H]1N6CCCCC6
Tekniske data
Kemisk navn eller materiale N-Desmethyl Vecuronium
Mængde 5 mg
InChI formel InChI=1S/C33H54N2O4/c1-22(36)38-30-19-24-11-12-25-26(33(24,4)21-29(30)35-17-9-6-10-18-35)13-14-32(3)27(25)20-28(31(32)39-23(2)37)34-15-7-5-8-16-34/h24-31H,5-21H2,1-4H3/t24-,25+,26-,27-,28-,29-,30-,31-,32-,33-/m0/s1
Formel vægt 542.4084