Få mere at vide
N-Desmethyl Ulipristal Acetate, TRC
Artikelnummer. 30136603
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136603 25 mg
30005293 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136603 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29600525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 159681-66-0
Molekylær formel C29 H35 N O4
Molekylvægt (g/mol) 461.59
Synonym 19-Norpregna-4,9-diene-3,20-dione, 17-(acetyloxy)-11-[4-(methylamino)phenyl]-, (11β)-, (11β)-17-(Acetyloxy)-11-[4-(methylamino)phenyl]-19-norpregna-4,9-diene-3,20-dione, CDB 3877, N-Monodemethyl CDB 2914, N-Desmethyl Ulipristal Acetate, N-Desmethylulipristal acetate, 17α-Acetoxy-11β-[4-(N-methylamino)-phenyl]-19-norpregna-4,9-diene-3,20-dione
IUPAC navn [(8S,11R,13S,14S,17R)-17-acetyl-13-methyl-11-[4-(methylamino)phenyl]-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
SMIL CNc1ccc(cc1)[C@H]2C[C@@]3(C)[C@@H](CC[C@]3(OC(=O)C)C(=O)C)[C@@H]4CCC5=CC(=O)CCC5=C24
Tekniske data
Kemisk navn eller materiale N-Desmethyl Ulipristal Acetate
Mængde 25 mg
InChI formel InChI=1S/C29H35NO4/c1-17(31)29(34-18(2)32)14-13-26-24-11-7-20-15-22(33)10-12-23(20)27(24)25(16-28(26,29)3)19-5-8-21(30-4)9-6-19/h5-6,8-9,15,24-26,30H,7,10-14,16H2,1-4H3/t24-,25+,26-,28-,29-/m0/s1
Formel vægt 461.2566