Få mere at vide
N-Desmethyl Trifluoperazine Dihydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Desmethyltrifluoperazine Dihydrochloride |
| CAS | 4617-82-7 |
| Molekylær formel | C20 H22 F3 N3 S . 2 Cl H |
| Mængde | 25 mg |
| Synonym | 10H-Phenothiazine, 10-[3-(1-piperazinyl)propyl]-2-(trifluoromethyl)-, hydrochloride (1:2), Phenothiazine, 10-[3-(1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (6CI,8CI), Desmethyltrifluoperazine Dihydrochloride |
| InChI formel | InChI=1S/C20H22F3N3S.2ClH/c21-20(22,23)15-6-7-19-17(14-15)26(16-4-1-2-5-18(16)27-19)11-3-10-25-12-8-24-9-13-25;;/h1-2,4-7,14,24H,3,8-13H2;2*1H |
| SMIL | Cl.Cl.FC(F)(F)c1ccc2Sc3ccccc3N(CCCN4CCNCC4)c2c1 |
| IUPAC navn | 10-(3-piperazin-1-ylpropyl)-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| Molekylvægt (g/mol) | 466.39 |
| Formel vægt | 465.102 |