Få mere at vide
N-Desmethyl Topotecan-d3 (>90%), TRC
Artikelnummer. 30353039
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30353039 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30353039 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2944021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale N-Desmethyl Topotecan-d3
CAS 1217633-79-8
Molekylær formel C22H18D3N3O5
Mængde 1 mg
InChI formel InChI=1S/C22H21N3O5/c1-3-22(29)15-7-17-19-11(9-25(17)20(27)14(15)10-30-21(22)28)6-12-13(8-23-2)18(26)5-4-16(12)24-19/h4-7,23,26,29H,3,8-10H2,1-2H3/t22-/m0/s1/i2D3
SMIL [2H]C([2H])([2H])NCc1c(O)ccc2nc3C4=CC5=C(COC(=O)[C@]5(O)CC)C(=O)N4Cc3cc12
Molekylvægt (g/mol) 410.44