Få mere at vide
N-Desmethyl Rosiglitazone-d4, TRC
Artikelnummer. 30367991
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367991 1 mg
30349476 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30367991 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2921671MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1215407-67-2
Molekylær formel C17H13D4N3O3S
Molekylvægt (g/mol) 347.42
Synonym 5-[[4-[2-(2-Pyridinylamino)ethoxy]phenyl]methyl]-2,4-thiazolidinedione-d4, SB 237216-d4
IUPAC navn 4-hydroxy-5-[[2,3,5,6-tetradeuterio-4-[2-(pyridin-2-ylamino)ethoxy]phenyl]methyl]-5H-1,3-thiazol-2-one
SMIL [2H]c1c([2H])c(OCCNc2ccccn2)c([2H])c([2H])c1CC3SC(=O)N=C3O
Tekniske data
Kemisk navn eller materiale N-Desmethyl Rosiglitazone-d4
Mængde 1 mg
InChI formel InChI=1S/C17H17N3O3S/c21-16-14(24-17(22)20-16)11-12-4-6-13(7-5-12)23-10-9-19-15-3-1-2-8-18-15/h1-8,14H,9-11H2,(H,18,19)(H,20,21,22)/i4D,5D,6D,7D
Formel vægt 347.1242