Få mere at vide
N-Desmethyl Propamocarb, TRC
Artikelnummer. 30012618
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012618 25 mg
17992745 100 mg
17922745 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012618 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29241025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1392231-41-2
Molekylær formel C8 H18 N2 O2
Molekylvægt (g/mol) 174.24
Synonym Carbamic acid, N-[3-(methylamino)propyl]-, propyl ester
IUPAC navn propyl N-[3-(methylamino)propyl]carbamate
SMIL CCCOC(=O)NCCCNC
Tekniske data
Kemisk navn eller materiale Propamocarb-N-desmethyl
Renhedsgrad noter GC
InChI formel InChI=1S/C8H18N2O2/c1-3-7-12-8(11)10-6-4-5-9-2/h9H,3-7H2,1-2H3,(H,10,11)
Mængde 25 mg
Formel vægt 174.1368
Procent renhed >95%