Få mere at vide
N-Desmethyl Prochlorperazine Dimaleate, TRC
Artikelnummer. 30368523
Skift visning
Mængde:
1 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30368523 2.5 mg 2.5mg
30365924 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30368523 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2920882.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13500-92-0
Molekylær formel C19 H22 Cl N3 S . 2 C4 H4 O4
Molekylvægt (g/mol) 592.06
Synonym 2-Chloro-10-[3-(1-piperazinyl)propyl]phenothiazine Maleate (1:2) (8CI), N-Desmethylprochlorperazine Dimaleate
IUPAC navn (Z)-but-2-enedioic acid;2-chloro-10-(3-piperazin-1-ylpropyl)phenothiazine
SMIL OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O.Clc1ccc2Sc3ccccc3N(CCCN4CCNCC4)c2c1
Tekniske data
Kemisk navn eller materiale N-Desmethyl Prochlorperazine Dimaleate
Mængde 2.5 mg
InChI formel InChI=1S/C19H22ClN3S.2C4H4O4/c20-15-6-7-19-17(14-15)23(16-4-1-2-5-18(16)24-19)11-3-10-22-12-8-21-9-13-22;2*5-3(6)1-2-4(7)8/h1-2,4-7,14,21H,3,8-13H2;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Formel vægt 591.1442